missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Mpro Tracer

Product Code. 30130410
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30130410 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30130410 Supplier Tocris Bioscience™ Supplier No. 8126/50U

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

SARS-CoV-2 main protease fluorescent ligand; TR-FRET acceptor

Mpro Tracer is a high affinity (Kd = 40 nM) non-covalent SARS-CoV-2 main protease fluorescent ligand. Mpro Tracer can be used in fluorescent polarisation (FP) assay for inhibitor screening/characterisation. It can be used as a FRET acceptor in time-resolved fluorescence resonance energy-transfer (TR-FRET) experiments. Excitation and emission maxima (λ) are 492 nm and 518 nm.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 50 μg
Target Fluorescent Ligands
Purity 0.95
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.