missing translation for 'onlineSavingsMsg'
Learn More

MPZL2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

145.00€ - 410.00€

Specifications

Antigen MPZL2
Concentration 0.31 mg/mL
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16839093
View Documents
Proteintech
11787-1-AP-20UL
20 μL
145.00€
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16829093
View Documents
Proteintech
11787-1-AP-150UL
150 μL
410.00€
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This antibody can be used to detect endogenous MPZL2, however, the reason for the additional band of 32 kDa is still unknown.

Epithelial V-like antigen (EVA) is expressed in thymus epithelium and strongly downregulated by thymocyte developmental progression. This gene is expressed in the thymus and in several epithelial structures early in embryogenesis.
Specifications

Specifications

MPZL2
Western Blot
Unconjugated
Rabbit
Human, Mouse, Rat
O60487, O70255
10205, 14012, 300679
MPZL2 Fusion Protein Ag2419
Primary
-20°C
MPZL2
0.31 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Epithelial V like antigen 1, EVA, EVA1, MPZL2, myelin protein zero like 2
MPZL2
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.