missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRGX3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-68959
This item is not returnable.
View return policy
Description
MRGX3 Polyclonal antibody specifically detects MRGX3 in Human samples. It is validated for Western Blot
Specifications
| MRGX3 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose | |
| G protein-coupled receptor MRGX3, G protein-coupled receptor SNSR2, GPCR, mas-related GPCR member X3, MAS-related GPR, member X3, mas-related G-protein coupled receptor member X3, MRGX3G protein-coupled receptor SNSR1, Sensory neuron-specific G-protein coupled receptor 1/2, SNSR1, SNSR2 | |
| Synthetic peptides corresponding to MRGPRX3 (MAS-related GPR, member X3) The peptide sequence was selected form the C terminal of MRGPRX3. Peptide sequence LDWKVLFCHVHLVSIFLSALNSSANPIIYFFVGSFRQRQNRQNLKLVLQR. The peptide sequence for this immunogen was taken from within the described region. | |
| 100 μg | |
| GPCR | |
| 117195 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| 0.5 mg/mL | |
| Western Blot 1.0 μg/mL | |
| Q96LA9 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction