missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MRPL39 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550062
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total MRPL39 protein.
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein. Two transcript variants encoding distinct isoforms have been described. A pseudogene corresponding to this gene is found on chromosome 5q.
Specifications
| MRPL39 | |
| Polyclonal | |
| Unconjugated | |
| MRPL39 | |
| 39S ribosomal protein L39, mitochondrial; 39S ribosomal protein L5, mitochondrial; C21orf8; C21orf92; L39mt; L5mt; Mitochondrial large ribosomal subunit protein mL39; mitochondrial ribosomal protein L39; MRPL39; MRP-L39; MRPL5; MRP-L5; MSTP003; ORF22; PRED22; PRED66; ribosomal protein, mitochondrial, L5; RPML5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 27393, 54148 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9JKF7, Q9NYK5 | |
| MRPL39 | |
| Synthesized peptide derived from internal of human MRPL39. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction