missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MRPS36 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584310
This item is not returnable.
View return policy
Description
Immunogen sequence: MMGSKMASAS RVVQVVKPHT PLIRFPDRRD NPKPNVSEAL RSAGLPSHS Highest antigen sequence indentity to the following orthologs: Mouse - 78%, Rat - 78%.
ROD1 is a functional homolog of nrd1, an S. pombe RNA-binding protein that suppresses the onset of differentiation.
Specifications
| MRPS36 | |
| Polyclonal | |
| Unconjugated | |
| MRPS36 | |
| 1110018B13Rik; 28S ribosomal protein S36, mitochondrial; DC47; mitochondrial ribosomal protein S36; Mrps36; MRP-S36; RT36; S36mt | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 92259 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Immunofluorescence | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P82909 | |
| MRPS36 | |
| Recombinant protein corresponding to Human MRPS36. Recombinant protein control fragment (Product #RP-106731). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction