missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MS4A6A Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand: Novus Biologicals H00064231-B01P
This item is not returnable.
View return policy
Description
MS4A6A Polyclonal antibody specifically detects MS4A6A in Human samples. It is validated for Western Blot
Specifications
| MS4A6A | |
| Polyclonal | |
| Western Blot 1:500 | |
| AAH22854 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.4) | |
| 4SPAN3, CD20 antigen-like 3, CD20L3MS4A6A-polymorph, CD20-like precusor, Four-span transmembrane protein 3, four-span transmembrane protein 3.1,4SPAN3.2, four-span transmembrane protein 3.2, HAIRB-iso, membrane-spanning 4-domains subfamily A member 6A, membrane-spanning 4-domains, subfamily A, member 6A, MGC131944, MGC22650, MS4A6MST090, MSTP090 | |
| MS4A6A (AAH22854, 1 a.a. - 248 a.a.) full-length human protein. MTSQPVPNETIIVPPSNVINFSQAEKPEPTNQGQDSLKKHLHAEIKVIGTIQILCGMMVLSLGIILASASFSPNFTQVTSTLLNSAYPFIGPFFFIISGSLSIATEKRLTKLLVHSSLVGSILSALSALVGFIILSVKQATLNPASLQCELDKNNIPTRSYVSYFYHDSLYTTDCYTAKASLAGSLSLMLICTLLEFCLAVLTAVLRWKQAYSDFPGSVLFLPHSYIGNSGMSSKMTHDCGYEELLTS | |
| 0.05 mg | |
| Signal Transduction | |
| 64231 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction