Learn More
MSH6 Antibody (CL13844) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-24582JF525
Product Code. 30297868
  • 737.00€ / 0.10mL
Description
MSH6 Monoclonal antibody specifically detects MSH6 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MSH6
Monoclonal
Janelia Fluor 525
DNA mismatch repair protein Msh6, G/T mismatch-binding protein, GTBPHNPCC5, GTMBP, hMSH6, HSAP, mutS (E. coli) homolog 6, mutS homolog 6 (E. coli), MutS-alpha 160 kDa subunit, p160, sperm-associated protein
This antibody was developed using a synthetic peptide derived from P52701, with the exact immunogen sequence remaining proprietary.
0.1 mL
Breast Cancer, Cancer, DNA Repair, Mismatch Repair
2956
Store at 4⪚C in the dark.
IgG2b
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
CL13844
50mM Sodium Borate
Mouse
Protein A purified