missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTHFR Mouse anti-Human, Clone: 1F7A3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66612-1-IG-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene catalyzes the conversion of 5,10-methylenetetrahydrofolate to 5-methyltetrahydrofolate, a co-substrate for homocysteine remethylation to methionine. Genetic variation in this gene influences susceptibility to occlusive vascular disease, neural tube defects, colon cancer and acute leukemia, and mutations in this gene are associated with methylenetetrahydrofolate reductase deficiency.Specifications
| MTHFR | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P42898 | |
| MTHFR | |
| MTHFR Fusion Protein Ag14970 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1F7A3 | |
| Unconjugated | |
| MTHFR | |
| MTHFR | |
| Mouse | |
| Protein A | |
| RUO | |
| 4524 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction