missing translation for 'onlineSavingsMsg'
Learn More

Methyltriethoxysilane, 98+%

Product Code. p-169457 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 mL
250 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
250mL
25mL
This item is not returnable. View return policy

Product Code. 10276180

Brand: Thermo Scientific Acros 174620250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

CAS: 2031-67-6 | C7H18O3Si | 178.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2031-67-6
Molecular Formula C7H18O3Si
Molecular Weight (g/mol) 178.30
MDL Number MFCD00009066
InChI Key CPUDPFPXCZDNGI-UHFFFAOYSA-N
Synonym methyltriethoxysilane, triethoxy methyl silane, methyl triethoxysilane, silane, triethoxymethyl, triethoxysilylmethane, union carbide a-162, ici-ep 5850, silane, methyltriethoxy, methaneorthosiliconic acid, triethyl ester, mtes
PubChem CID 16241
IUPAC Name triethoxy(methyl)silane
SMILES CCO[Si](C)(OCC)OCC

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point -40.0°C
Color Colorless
Density 0.8900g/mL
Boiling Point 141.0°C to 143.0°C
Flash Point 23°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula CH3Si(OC2H5)3
Refractive Index 1.3825 to 1.3845
Quantity 25 mL
Beilstein 04,629
Specific Gravity 0.89
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents
Viscosity 0.6 mPa.s (20°C)
Formula Weight 178.3
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material Methyltriethoxysilane, 98+%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.