Learn More
MTMR14 Monoclonal Antibody (OTI7B2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA501737S
Description
MTMR14 Monoclonal antibody specifically detects MTMR14 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| MTMR14 | |
| Monoclonal | |
| 0.43 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8NEX9 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 121214 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, Western Blot | |
| OTI7B2 | |
| Unconjugated | |
| SDR9C7 | |
| 1110061O04Rik; AW553738; C3orf29; C76151; egg-derived tyrosine phosphatase homolog; fc14d11; HCV NS5A-transactivated protein 4 splice variant A-binding protein 1; hJumpy; jumpy; mJumpy; Mtmr14; myotubularin related protein 14; myotubularin-related protein 14; NS5ATP4ABP1; RGD1304842; wu:fc14d11 | |
| Full length human recombinant protein of human SDR9C7 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.