missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTR Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
198.00€ - 468.00€
Specifications
| Antigen | MTR |
|---|---|
| Dilution | Western Blot 1:200-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
MTR Polyclonal antibody specifically detects MTR in Human, Mouse samples. It is validated for Western BlotSpecifications
| MTR | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Lipid and Metabolism | |
| PBS (pH 7.3), 50% glycerol | |
| 4548 | |
| IgG | |
| Affinity purified |
| Western Blot 1:200-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| 5-methyltetrahydrofolate-homocysteine methyltransferase, 5-methyltetrahydrofolate-homocysteine methyltransferase 1, cblG, cobalamin-dependent methionine synthase, EC 2.1.1.13, FLJ33168, FLJ43216,5-methyltetrahydrofolate--homocysteine methyltransferase, FLJ45386, methionine synthase, MS, Vitamin-B12 dependent methionine synthase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1056-1265 of human MTR (NP_000245.2). AEPIATFYGLRQQAEKDSASTEPYYCLSDFIAPLHSGIRDYLGLFAVACFGVEELSKAYEDDGDDYSSIMVKALGDRLAEAFAEELHERVRRELWAYCGSEQLDVADLRRLRYKGIRPAPGYPSQPDHTEKLTMWRLADIEQSTGIRLTESLAMAPASAVSGLYFSNLKSKYFAVGKISKDQVEDYALRKNISVAEVEKWLGPILGYDTD | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title