Learn More
MUC1 Antibody (139H2), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-47888MFV500
Product Code. 18789762
  • 793.00€ / 0.10mL
Description
MUC1 Monoclonal antibody specifically detects MUC1 in Human, Mouse samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MUC1
Monoclonal
mFluor Violet 500 SE
50mM Sodium Borate
Mouse
Protein A or G purified
RUO
Primary
Human, Mouse
Purified
Flow Cytometry
139H2
Flow Cytometry
Breast carcinoma-associated antigen DF3, Carcinoma-associated mucin, CD227, CD227 antigen, DF3 antigen, EMA, episialin, H23 antigen, H23AG, KL-6, MAM6, MUC-1, MUC1/ZD, mucin 1, cell surface associated, mucin 1, transmembrane, mucin-1, Peanut-reactive urinary mucin, PEMMUC-1/SEC, PEMT, Polymorphic epithelial mucin, PUMMUC-1/X, tumor associated epithelial mucin, Tumor-associated epithelial membrane antigen, Tumor-associated mucin
Human milk-fat globule membranes (HMFGM) (Uniprot: P15941 )
0.1 mL
Cancer, Cellular Markers, Extracellular Matrix, Inflammation, Signal Transduction
4582
Store at 4C in the dark.
IgG1 κ