Learn More
MUC1 Antibody (CL8733) [Alexa Fluor™ 700], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-05503AF700
Product Code. 30292079
  • 737.00€ / 0.10mL
Description
MUC1 Monoclonal antibody specifically detects MUC1 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MUC1
Monoclonal
Alexa Fluor 700
Breast carcinoma-associated antigen DF3, Carcinoma-associated mucin, CD227, CD227 antigen, DF3 antigen, EMA, episialin, H23 antigen, H23AG, KL-6, MAM6, MUC-1, MUC1/ZD, mucin 1, cell surface associated, mucin 1, transmembrane, mucin-1, Peanut-reactive urinary mucin, PEMMUC-1/SEC, PEMT, Polymorphic epithelial mucin, PUMMUC-1/X, tumor associated epithelial mucin, Tumor-associated epithelial membrane antigen, Tumor-associated mucin
This antibody was developed using a recombinant protein derived from P15941, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cancer, Cellular Markers, Extracellular Matrix, Inflammation, Signal Transduction
4582
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Immunohistochemistry (Paraffin)
CL8733
50mM Sodium Borate
Mouse
Protein A purified