Learn More
MUC1 (CD227) Mouse, anti-Human, PE, Clone: 16A, STEMCELL Technologies
Mouse Monoclonal Antibody
Brand: Stemcell Technologies 60155PE
Description
- The 16A antibody reacts with human MUC1, a large (> 250 kDa) heavily glycosylated type 1 transmembrane protein expressed on the surface of most glandular and ductal epithelial cells and a variety of hematopoietic cells
- A characteristic feature of the MUC1 glycoprotein is a core domain composed of a variable number of tandem repeats and multiple oligosaccharide side chains
- Because the extracellular portion of MUC1 can extend beyond most cell surface proteins, it is thought to play a role in cell-cell and cell-substrate adhesion
- The protein is highly expressed by a majority of human adenocarcinomas and is associated with a poor prognosis
- In the mammary gland, MUC1 is localized on the apical plasma membrane of luminal epithelial cells
- The clone 16A antibody has a higher affinity for the glycosylated form of MUC1
Specifications
| MUC1 (CD227) | |
| Monoclonal | |
| PE | |
| 100 Tests | |
| Human | |
| Airway Cells |
| Immunohistochemistry, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| 16A | |
| Mouse | |
| Primary | |
| IgG1 λ |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.