missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MUC4 Antibody (781631), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB8195H
This item is not returnable.
View return policy
Description
MUC4 Monoclonal antibody specifically detects MUC4 in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| MUC4 | |
| Monoclonal | |
| HRP | |
| PBS | |
| Mouse | |
| 0.1 mL | |
| Cancer | |
| 4585 | |
| Store at 4C in the dark. |
| Flow Cytometry, CyTOF | |
| 781631 | |
| Flow Cytometry, CyTOF-ready | |
| Ascites sialoglycoprotein, ASGP, HSA276359, MUC-4, mucin 4, cell surface associated, mucin 4, tracheobronchial, mucin-4, Pancreatic adenocarcinoma mucin, Testis mucin, Tracheobronchial mucin | |
| E. coli -derived recombinant human MUC4, Pro1072-Ser1317, Accession # Q99102 | |
| RUO | |
| Primary | |
| Human | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction