missing translation for 'onlineSavingsMsg'
Learn More

4-(Dimethylamino)-1,1-dimethoxybut-3-en-2-one, 97%, Thermo Scientific™

Product Code. 10452454
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
50g
This item is not returnable. View return policy

Product Code. 10452454

Brand: Thermo Scientific Maybridge RF03845EE

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 67751-23-9
Molecular Formula C8H15NO3
Molecular Weight (g/mol) 173.212
InChI Key DFZIBCAWOSFLFR-AATRIKPKSA-N
Synonym 4-dimethylamino-1,1-dimethoxybut-3-en-2-one, e-4-dimethylamino-1,1-dimethoxybut-3-en-2-one, 1,1-dimethoxy-4-dimethylaminobut-3-en-2-one, 1e-4,4-dimethoxy-3-oxobut-1-en-1-yl dimethylamine, 3e-4-dimethylamino-1,1-dimethoxybut-3-en-2-one, e-1,1-dimethoxy-4-dimethylaminobut-3-en-2-one, hcvl@nbfryyuffjjafp, e-4-dimethylamino-1,1-dimethoxy-but-3-en-2, 4-dimethylamino-1,1-dimethoxy-3-butene-2-one, e-4-dimethylamino-1,1-dimethoxy-but-3-en-2-one
PubChem CID 5709580
IUPAC Name (E)-4-(dimethylamino)-1,1-dimethoxybut-3-en-2-one
SMILES CN(C)C=CC(=O)C(OC)OC

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula (CH3)2NCH=CHC(O)CH(OCH3)2
Quantity 50 g
Formula Weight 173.21
Percent Purity 97%
Chemical Name or Material 4-(Dimethylamino)-1, 1-dimethoxybut-3-en-2-one
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.