Learn More
Heidolph™ Multi-channel Pump Head
Product Code. 10504932 Shop All Heidolph Instruments Products
Change view
Click to view available options
Model:
C 12
C 4
C 8
Holds:
12 small cassettes
4 small cassettes
8 medium cassettes
Includes:
4 rollers
8 rollers
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model Holds Includes Unit Size
10504932 C 12 12 small cassettes 8 rollers Each
10448401 C 4 4 small cassettes 8 rollers Each
10504742 C 8 8 medium cassettes 4 rollers Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10504932 Supplier Heidolph™ Supplier No. 5248122000
  • ONLINE EXCLUSIVE PRICE
    4360.00€
    3743.76€ / Pack of 1
    Save 616.24€
    14% Off
Estimated Shipment: 10-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Increase your throughput by running more cassettes on one single pump drive and pump head system
  • Separate metering into multiple vessels with different feed rates in a single operation by using different tubing dimensions to adjust flow rate
  • Save time - tubing change is done in just seconds
  • Change your cassettes easily, even during operation – there are no restrictions
  • Cassette adjustments and changes are simple, just click in place
  • No additional purchase of a pump drive needed: upgrade your single-channel pump drive with just one adaptor for multi-channel use and pick the head/cassette configuration that matches your application needs
TRUSTED_SUSTAINABILITY
Specifications
Holds 12 small cassettes
Includes 8 rollers
Model C 12
Type Pump Head