Learn More
FTM Technologies™ Multi-Operator Fume Hoods
Product Code. 11778054
Change view
Click to view available options
Includes:
4 extraction inputs
6 extraction inputs
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Unit Size
11778054 6 extraction inputs Each
11778044 4 extraction inputs Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Product Code. 11778054 Supplier FTM Technologies™ Supplier No. HI4P6S
  • ONLINE EXCLUSIVE PRICE
    1620.00€
    1326.49€ / Pack of 1
    Save 293.51€
    18% Off
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Used for filtration where extraction arms or other accessories are needed. FTM Technologies™ Multi-Operator Fume Hoods provide multi-workstation filtration for welding fumes, solvents and other hazardous substances.
  • Available with either 4 or 6 extraction inputs
  • Delivered with ringed PVC pipe
  • Compatible with the use of prefilters
  • Can be used as a recirculating (ductless) fume hood, using active carbon filtration or dust filter, or with an external output (external filter plus exhaust duct)
Specifications
Dimensions (L x W x H) 350 x 200 x 430 mm
Flow Rate 50 to 450 m3/h
Includes 6 extraction inputs
Voltage 230 V
Weight (Metric) 6 kg
Frequency 50 Hz