Learn More
Mecmesin™ Multi test Intelligent Loadcell
Product Code. 11923858
Encompass
Change view
Click to view available options
Multi test Intelligent Loadcell
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11923858 Multi test Intelligent Loadcell
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
R\u00E9cupel-bijdrage uitgesloten
Product Code. 11923858 Supplier Mecmesin™ Supplier No. 879005
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
R\u00E9cupel-bijdrage uitgesloten
Compatible with all test stands of equal or higher capacity than load cell.
  • Transducer which performs functionality of converting force into an electric output
  • Capacity: 250N
  • Overall daylight reduction: 15(0.59)
  • Dimension (D x W x H): 46.5 x 72 x 48cm
Specifications
Product Type Loadcell
Description Capacity: 25N
For Use With (Application) Transducer: converts force into an electric output which can be measured