Learn More
ATAGO™ Multi-Wavelength Abbe Refractometer DR-M2
Product Code. 11446248
Change view
Click to view available options
Dimensions (L x W x H):
130 x 290 x 310 mm (Refractometer), 150 x 330 x 110 mm (Light Source Unit)
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) Unit Size
11446248 130 x 290 x 310 mm (Refractometer), 150 x 330 x 110 mm (Light Source Unit) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Product Code. 11446248 Supplier ATAGO™ Supplier No. 1410
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
The Multi-Wavelength Abbe Refractometer DR-M2 digitally displays the measurement result of refractive index or Abbe number on the LCD.
  • Refractive index or Abbe number (νd or νe) can be measured at different wavelengths ranging from 450 to 1100 nm.
  • The DR-M2 digitally displays measurement result of refractive index or Abbe number on the LCD.
  • Measurement can be achieved by matching the boundary line with the intersection point of the cross hairs.
  • This refractometer is connectable with Digital printer DP-63 (B) (Optional).
Specifications
Type Benchtop Refractometer
Refractive Index 1.3278 to 1.7379 (at 450 nm), 1.3000 to 1.7100 (at 589 nm), 1.2912 to 1.7011 (at 680 nm), 1.2743 to 1.6840 (at 1100 nm)
Accuracy ±0.0002
Display LCD
Weight (Metric) 6.0 kg (Refractometer), 3.2 kg (Light Source Unit)
Dimensions (L x W x H) 130 x 290 x 310 mm (Refractometer), 150 x 330 x 110 mm (Light Source Unit)
Temperature Range (Metric) 5°C to 50°C
Electrical Requirements AC100
Voltage 100/240 V
Frequency 50/60 Hz