Learn More
Mecmesin™ Multifunction Force/Torque Indicator
Product Code. 11947979
Encompass
Change view
Click to view available options
Multifunction Force/Torque Indicator
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11947979 Multifunction Force/Torque Indicator
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
R\u00E9cupel-bijdrage uitgesloten
Product Code. 11947979 Supplier Mecmesin™ Supplier No. 851401
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
R\u00E9cupel-bijdrage uitgesloten
Features high speed sampling rate.
  • Advanced indicator with precision external force, torque sensors
  • High-speed 5000Hz sampling rate
  • Audio/visual alarms
  • RS-232 interface
  • Graphical display load-bar graph
  • Live and peak readings simultaneously
  • Takes high-accuracy measurements with precision external smart transducers
  • Meter automatically recognizes transducers—no calibration or setup
  • Large, backlit display provides graphical representation
  • Sampling rate: 5000 HZ
  • Data storage: 500
  • Output: RS-232
Specifications
Product Type Indicator
Description Multifunction force/torque indicator
For Use With (Application) Takes high-accuracy measurements with precision external smart transducer