Learn More
Horiba™ Multiparameter Water Quality Meter Kit
Product Code. 11958128
Encompass
Change view
Click to view available options
Range:
Turbidity: 0 to 1000NTU
Turbidity: 0 to 800NTU
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Range unitSize
11958128 Turbidity: 0 to 800NTU Each
11998128 Turbidity: 0 to 1000NTU Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Product Code. 11958128 Supplier Horiba™ Supplier No. U52G10M
This item requires a price update prior to being ordered.
Request a Price for this item here.
, 02 950 59 478
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Tough and powerfully designed multiparameter water tester. Horiba™ Multiparameter Water Quality Meter Kit simultaneously measures and indicates results for up to 11 parameters.
  • Auto-calibration feature provides hassle free calibration of pH, dissolved oxygen, conductivity, turbidity and depth.
  • Shock-resistant cover designed for rough treatment in the field and is easily cleaned.
  • Cable can be easily connected and disconnected with quick-connect fitting.
  • Auto hold function freezes average data values on the screen to offer more time to verify or transcribe data.
  • Diagnostic functions notify the user of errors.
  • Integral USB connection for data transfer to a PC. USA cable is sold separately and includes software.
  • Selectable measurement units allow the operator to report data without the need to convert data to desired units of measure.
  • Multiple sensors allow for the measurement of 11 parameters simultaneously (pH, pH(mv), ORP, DO, COND, Salinity, TDS, Seawater Specific Gravity, Temperature, Turbidity, Water Depth).
  • Turbidity sensor U-53 conforms to US EPA method 180.1.
  • Precision has been improved over conventional instruments.
  • Replaceable turbidity sensor with wiper has a resolution of 0.01 NTU.
  • Turbidity sensor of U-54 conforms to EN ISO 7027. The model U-54 has a resolution of 0.01 NTU.
  • Improved stability of the dissolved oxygen sensor has been achieved with a new 3 electrode design for fast response and polarographic sensor for ease of maintenance.
  • pH and ORP electrodes can be replaced individually to reduce replacement costs
Specifications
Accuracy Turbidity: ±5% or ±1NTU, whichever is greater.
Outputs USB
Accuracy (Conductivity) ±1% F.S.
Accuracy (DO) 0 to 20mg/L: ±0.2mg/L20 to 50mg/L: ±0.5mg/L
Accuracy (pH) ±0.1pH
Accuracy (Salinity) ±3 PPT
Accuracy (TDS) ±5g/L
Backlight Option Yes
Battery Life ∼70 hr. (without backlight)
Battery Type 4 C Alkaline
Display Type LCD, 320 X 240, Backlit (black and white)
Range (Conductivity) 0 to 100 mS/cm
Range (pH) 0 to 14
Range (Salinity) 0 to 70 PPT (permillage)
Range (TDS) 0 to 100 g/L
Resolution (pH) 0.01
Temperature Range (Metric) -10°C to +55°C
Includes Analyzer and Probe
Memory 10,000
Methods Turbidity: LED light source and 30° scattering method.
Product Type Portable multiparameter water quality meter.
Range Turbidity: 0 to 800NTU
Range (ORP) −2000mV - +2000mV
Resolution Turbidity: 0 to 99.9NTU: 0.1, 100 to 800NTU: 1.0
Resolution (Conductivity) 0.000 to 0.999mS/cm:0.001 1.00 to 9.99mS/cm: 0.0110.0 to 99.9mS/cm: 0.10.0 to 99.9mS/m: 0.10.100 to 0.999S/m: 0.0011.00 to 9.99S/m: 0.01
Resolution (RDO) 0.01mg/L
Resolution (Salinity) 0.1 PPT
Resolution (TDS) 0.1% F.S.
Salinity Measurement Modes Conductivity conversion
Temperature Accuracy JIS class B platinum thermometer sensor (±0.3+0.005 /t/)
Temperature Resolution 0.01°C
Length (Metric) Probe: 340 mm, Cable: 2 m standard, 10 m, 30 m options
Width (Metric) 96 mm probe
Weight (Metric) 1800 g