missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MUM1 Antibody (4G10), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-47396-0.025ml
This item is not returnable.
View return policy
Description
MUM1 Monoclonal specifically detects MUM1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| MUM1 | |
| Monoclonal | |
| Unconjugated | |
| EXPAND1, FLJ14868, FLJ22283, melanoma associated antigen (mutated) 1, melanoma ubiquitous mutated protein, MGC131891, MGC163315, MUM-1HSPC211, Mutated melanoma-associated antigen 1, Protein expandere, PWWP domain-containing protein MUM1 | |
| Mouse | |
| 78.6 kDa | |
| 0.025 mL | |
| Melanoma Cell Markers | |
| 84939 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
| Western Blot | |
| 4G10 | |
| Western Blot 1:500-1:2000, ELISA 1:10000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| MUM1 | |
| Purified recombinant fragment of human MUM1 expressed in E. Coli. | |
| Unpurified | |
| RUO | |
| Primary | |
| Human | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction