missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Musashi-1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-32812
This item is not returnable.
View return policy
Description
Musashi-1 Polyclonal specifically detects Musashi-1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen.
Specifications
| Musashi-1 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
| MSI1 | |
| Recombinant protein encompassing a sequence within the center region of human Musashi 1. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Tested in human. Not yet tested in any other species. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen) | |
| 0.69mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunoprecipitation, Immunohistochemistry-Frozen 1:100-1:1000 | |
| Musashi (Drosophila) homolog 1, musashi homolog 1 (Drosophila), Musashi-1, RNA-binding protein Musashi homolog 1 | |
| Rabbit | |
| 39 kDa | |
| 0.1 mL | |
| Cancer, Cellular Markers, Neurodegeneration, Neuronal Cell Markers, Neuronal Stem Cell Markers, Neuroscience, Stem Cell Markers, Stem Cells | |
| 4440 | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction