missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Muscarinic Acetylcholine Receptor M5/CHRM5 Antibody (7E5H11), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27131-0.1ml
This item is not returnable.
View return policy
Description
Muscarinic Acetylcholine Receptor M5/CHRM5 Monoclonal antibody specifically detects Muscarinic Acetylcholine Receptor M5/CHRM5 in Human samples. It is validated for Flow Cytometry,ELISA,Western Blot
Specifications
| Muscarinic Acetylcholine Receptor M5/CHRM5 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000 | |
| cholinergic receptor, muscarinic 5, HM5, MGC41838, muscarinic acetylcholine receptor M5 | |
| Purified recombinant fragment of human Muscarinic Acetylcholine Receptor M5/CHRM5 expressed in E. Coli. | |
| 0.1 mL | |
| GPCR, Neuroscience | |
| 1133 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
| Flow Cytometry, ELISA, Western Blot | |
| 7E5H11 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction