Learn More
Honeywell™ Mutex E Light Protective Clothing
Product Code. 15693346
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15693346 X-Large Each
15683346 Small Each
15673346 Medium Each
15663346 Large Each
15603356 2X-Large Each
15613356 3X-Large Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 15693346 Supplier Honeywell™ Supplier No. 4500101XL
  • ONLINE EXCLUSIVE PRICE
    11.05€
    6.66€ / Pack of 1
    Save 4.39€
    39% Off
Estimated Shipment: 16-04-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Breathable, single-use protective clothing designed to protect against particle contamination and chemical splashes.
  • Antistatic
  • Silicone and latex free
  • Excellent ergonomics and fitting
    • 3-piece hood
    • Special crotch design
    • Outside flap and dual slider sipper
    • Breathability
    • 3-layer polypropylene
TRUSTED_SUSTAINABILITY
Specifications
Product Type Chemical-resistant Coveralls
Size X-Large
Gender Unisex
Material 3-layer Polypropylene
Color White
Protection Class Protection type: 5, 6
Certifications/Compliance (Europe) Ec Category PPE: 3, EC Certificate: 78815277, EC Attestation: 78815277
Thickness 56 gsm