missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MVK Recombinant Rabbit Monoclonal Antibody (24GB115)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557477
This item is not returnable.
View return policy
Description
MVK Recombinant Monoclonal Antibody for Western Blot, ICC/IF
Mevalonate kinase (MVK) is an early enzyme in isoprenoid and sterol synthesis. Mevalonate kinase catalyzes the ATP-dependent phosphorylation of mevalonic acid to form mevalonate 5-phosphate. Deficiency in MVK activity contributes to mevalonic aciduria and hyperimmunoglobulinemia D/periodic fever syndrome (HIDS).
Specifications
| MVK | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Mvk | |
| 2310010A05Rik; AI256848; AI414037; KIME; LH receptor mRNA-binding protein; LHR mRNA binding protein; Lrbp; Mevalonate kinase; mevalonate kinase 1; MK; Mvk; MVLK; POROK3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 4598, 81727 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB115 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P17256, Q03426 | |
| Mvk | |
| A synthesized peptide derived from human MVK (aa 320-370). | |
| 20 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction