missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MX1 Polyclonal Antibody

Rabbit Polyclonal Antibody
208.00€ - 504.00€
Specifications
| Antigen | MX1 |
|---|---|
| Concentration | 1 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17740704
![]() |
Invitrogen™
PA5119298 |
200 μL |
504.00€
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17733154
|
Invitrogen™
PA5121755 |
20 μL |
208.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MX1 Polyclonal Antibody for Western Blot, IHC (P)
This gene encodes a guanosine triphosphate (GTP)-metabolizing protein that participates in the cellular antiviral response. The encoded protein is induced by type I and type II interferons and antagonizes the replication process of several different RNA and DNA viruses. There is a related gene located adjacent to this gene on chromosome 21, and there are multiple pseudogenes located in a cluster on chromosome 4. Alternative splicing results in multiple transcript variants.Specifications
| MX1 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human, Mouse | |
| Mx1 | |
| AI893580; GTP-binding protein; IFI78; IFI-78K; Influenza resistance protein; Interferon-induced GTP-binding protein Mx1; Interferon-induced GTP-binding protein Mx1, N-terminally processed; Interferon-induced protein p78; interferon-inducible myxovirus resistance-1 protein; interferon-inducible protein p78; interferon-regulated resistance GTP-binding protein MxA; Mx; MX dynamin like GTPase 1; MX dynamin-like GTPase 1; Mx1; Mx-1; Mx1B; Mx1-b; MxA; Myxoma resistance protein 1; myxovirus (influenza virus) resistance 1; myxovirus (influenza virus) resistance 1, interferon-inducible protein p78; myxovirus (influenza) resistance 1 polypeptide; myxovirus resistance protein 1 | |
| Mx1 | |
| Primary | |
| Antigen affinity chromatography, Protein A |
| 1 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P09922, P20591 | |
| 17857, 4599 | |
| Recombinant protein Myxovirus Resistance 1. The antigen corresponds to amino acid range 80-342 of the target protein. | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title