Learn More
Mycobacterium tuberculosis MPT64 Polyclonal Antibody
Brand: Thermo Scientific PA5117509
Specifications
| Mycobacterium tuberculosis MPT64 | |
| Polyclonal | |
| Unconjugated | |
| mpt64 | |
| mpt64 | |
| Recombinant Mycobacterium tuberculosis Immunogenic protein MPT64 protein (24-228aa). | |
| 100 μg | |
| Primary | |
| Bacteria | |
| Antibody | |
| IgG |
| Western Blot | |
| 2.6 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| P9WIN9 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 885925 | |
| -20°C or -80°C if preferred | |
| Liquid |