missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myelin PLP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 5 publications
Brand: Novus Biologicals NB100-74503-0.025ml
This item is not returnable.
View return policy
Description
Myelin PLP Polyclonal specifically detects Myelin PLP in Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| Myelin PLP | |
| Polyclonal | |
| Western Blot 1:1000, Immunocytochemistry/Immunofluorescence, Immunocytochemistry 1:1000 | |
| HLD1, lipophilin, major myelin proteolipid protein, MMPL, myelin proteolipid protein, PLP, PLP/DM20, PMD, proteolipid protein 1, spastic paraplegia 2, uncomplicated, SPG2 | |
| Rabbit | |
| Unpurified | |
| RUO | |
| Primary | |
| PLP | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| Whole antisera with 0.05% Sodium Azide | |
| PLP1 | |
| Synthetic peptide corresponding to residues C(109) G K G L S A T V T G G Q K G R G S R G(128) of PLP. | |
| 0.025 mL | |
| Immune System Diseases, Immunology | |
| 5354 | |
| Mouse | |
| Antisera |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction