Learn More
MYH1 Mouse anti-Human, Unconjugated, Clone: 1G10H9, Proteintech
Mouse Monoclonal Antibody
145.00€ - 410.00€
Specifications
| Antigen | MYH1 |
|---|---|
| Clone | 1G10H9 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, Immunohistochemistry, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16889845
|
Proteintech
67299-1-IG-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16879845
|
Proteintech
67299-1-IG-150UL |
150 μL |
410.00€
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The MYH1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human MYH1. This antibody recognizes human, mouse, pig antigen. The MYH1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| MYH1 | |
| 1000 μg/mL | |
| Monoclonal | |
| Mouse | |
| 4619 | |
| Primary |
| 1G10H9 | |
| Western Blot, Immunohistochemistry, ELISA | |
| Unconjugated | |
| Human, Pig, Mouse | |
| IgG2a |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.