missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYH7 Monoclonal Antibody (TH81 (BGN/04/4481)), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA183347
This item is not returnable.
View return policy
Beschreibung
Mouse anti Rabbit muscle myosin heavy chain antibody, clone TH81 recognizesthe heavy chain of rabbit muscle myosin.
Muscle myosin is a hexameric protein containing 2 heavy chain subunits, 2 alkali light chain subunits, and 2 regulatory light chain subunits. This gene encodes the beta (or slow) heavy chain subunit of cardiac myosin. It is expressed predominantly in normal human ventricle. It is also expressed in skeletal muscle tissues rich in slow-twitch type I muscle fibers. Changes in the relative abundance of this protein and the alpha (or fast) heavy subunit of cardiac myosin correlate with the contractile velocity of cardiac muscle. Its expression is also altered during thyroid hormone depletion and hemodynamic overloading. Mutations in this gene are associated with familial hypertrophic cardiomyopathy, myosin storage myopathy, dilated cardiomyopathy, and Laing early-onset distal myopathy.
Spezifikation
| MYH7 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with <0.1% sodium azide; pH 7.2 | |
| P02564, P04461, P12883, Q91Z83 | |
| Myh7 | |
| Myosin heavy chain from rabbit muscle. | |
| 200 μg | |
| Primary | |
| Human, Mouse, Rabbit, Rat | |
| Antibody | |
| IgG1 |
| ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| TH81 (BGN/04/4481) | |
| Unconjugated | |
| Myh7 | |
| beta cardiac myosin heavy chain; beta myosin heavy chain; beta-MHC; beta-myosin; beta-myosin heavy chain; beta-myosin-heavy-chain; B-MHC; cardiac muscle myosin heavy chain 7 beta; cardiac myosin heavy chain beta isoform; CMD1S; CMH1; LOC100727138; mg:cb02g011; MPD1; Myh7; MYH-beta/slow; MyHC-alpha; Myhcb; Myhc-b; myHC-beta; MyHC-I; MyHC-slow; myopathy, distal 1; myosin beta heavy chain; myosin heavy chain 1; myosin heavy chain 7; myosin heavy chain cardiac alpha; myosin heavy chain polypeptide 7 cardiac muscle fetal; myosin heavy chain slow; myosin heavy chain slow isoform; myosin heavy chain slow type 1 (beta cardiac); myosin heavy chain, cardiac muscle beta isoform; myosin heavy chain, cardiac muscle, fetal; myosin heavy chain, polypeptide 7; myosin, heavy chain 7, cardiac muscle, beta; myosin, heavy polypeptide 7, cardiac muscle, beta; myosin-7; rhabdomyosarcoma antigen MU-RMS-40.7A; slow myosin heavy chain; smyhc3; SPMD; SPMM; type slow/beta myosin heavy chain; Unknown (protein for IMAGE:7985957); ventricular myosin heavy chain; vmhc | |
| Mouse | |
| Protein G | |
| RUO | |
| 100101566, 140781, 29557, 4625 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur