missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYH9 Rabbit anti-Human, Mouse, Rat, Zebrafish, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11128-1-AP-20UL
This item is not returnable.
View return policy
Description
TMP21 is a ubiquitously expressed protein that is involved in vesicular targeting and protein transport. More recent experiments have shown that it is also a component in the presenilin complex and modulates the -gamma-secretase but not the epsilon-secretase cleavage activity of the amyloid precursor protein. The presenilin complex is composed of the proteins APH1, nicastrin, and PEN2 in addition to presenilin-1. Together, these proteins cleave the amyloid precursor protein at what is known as the -gamma- and epsilon-sites and can lead to the accumulation of the Abeta cleavage product that is associated with Alzheimer′s disease. Co-immunoprecipitation experiments using antibodies against these proteins also yielded TMP21 indicating that TMP21 may play a role in the regulation of this complex. Suppression of TMP21 expression by siRNA in transfected cells caused increased -gamma-secretase activity but not epsilon-secretase activity, and increased Abeta production, demonstrating that TMP21 can modulate -gamma-secretase activity.Specifications
| MYH9 | |
| Polyclonal | |
| Unconjugated | |
| MYH9 | |
| MYH9, Myosin 9, Myosin heavy chain 9, myosin IIa, Nonmuscle myosin IIa | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 100911597, 17886, 333938, 4627 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P35579, Q62812, Q8VDD5 | |
| Myh9, myh9a | |
| MYH9 Fusion Protein Ag1612 | |
| 20 μL | |
| Primary | |
| Rat, Human, Zebrafish, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction