missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOAT 2 Antibody (OTI2D3), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74271AF405
This item is not returnable.
View return policy
Description
SOAT 2 Monoclonal specifically detects SOAT 2 in Human samples. It is validated for Immunohistochemistry.
Specifications
| SOAT 2 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| SOAT2 | |
| Recombinant protein fragment corresponding to amino acids 1-120 of human SOAT 2 (NP_003569) produced in E.coli. | |
| 0.1 mL | |
| 8435.0 | |
| Store at 4°C in the dark. |
| Immunohistochemistry | |
| OTI2D3 | |
| Immunohistochemistry | |
| ACACT2MGC116732, ACAT-2, ACAT2ARGP2, acyl Co-A: cholesterol acyltransferase 2, acyl coenzyme A:cholesterol acyltransferase 2, acyl-CoA:cholesterol acyltransferase 2, Acyl-coenzyme A:cholesterol acyltransferase 2, Cholesterol acyltransferase 2, EC 2.3.1.26, sterol O-acyltransferase 2 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction