missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYL5 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14249-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes one of the myosin light chains, a component of the hexameric ATPase cellular motor protein myosin. Myosin is composed of two heavy chains, two nonphosphorylatable alkali light chains, and two phosphorylatable regulatory light chains. This gene product, one of the regulatory light chains, is expressed in fetal muscle and in adult retina, cerebellum, and basal ganglia.Specifications
| MYL5 | |
| Polyclonal | |
| Unconjugated | |
| MYL5 | |
| MYL5, MyLC 2, MYLC2, Myosin light chain 5 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 4636 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q02045 | |
| MYL5 | |
| MYL5 Fusion Protein Ag5522 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction