missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myofibroblast Marker Antibody (PR 2D3), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08768V
This item is not returnable.
View return policy
Description
Myofibroblast Marker Monoclonal antibody specifically detects Myofibroblast Marker in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
Specifications
| Myofibroblast Marker | |
| Monoclonal | |
| DyLight 405 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry (Paraffin) | |
| PR 2D3 | |
| 50mM Sodium Borate | |
| Crude homogenate of normal human colorectal mucosa | |
| 100 μg | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction