missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myosin heavy chain 2 Mouse anti-Human, Rat, Rabbit, Clone: TH81 (BGN/04/4481), Novus Biologicals™
Mouse Monoclonal Antibody
472.00€
Specifications
| Antigen | Myosin heavy chain 2 |
|---|---|
| Clone | TH81 (BGN/04/4481) |
| Concentration | 1 mg/mL |
| Dilution | ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 |
| Applications | ELISA |
Description
Myosin heavy chain 2 Monoclonal specifically detects Myosin heavy chain 2 in Human, Rat, Rabbit samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Myosin heavy chain 2 | |
| 1 mg/mL | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Cell Biology, Cellular Markers | |
| PBS (pH 7.2) | |
| 4620 | |
| IgG1 | |
| Protein A purified |
| TH81 (BGN/04/4481) | |
| ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| fast 2a myosin heavy chain, MYH2A, MYHas8, MyHC-2A, MyHC-IIa, MYHSA2, myosin heavy chain 2, myosin heavy chain 2a, myosin heavy chain IIa, myosin, heavy chain 2, skeletal muscle, adult, myosin, heavy polypeptide 2, skeletal muscle, adult, myosin-2, skeletal muscle, adult 2, type IIA myosin heavy chain | |
| Myosin heavy chain from rabbit muscle | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title