Learn More
myosin X Antibody (CL9032) [DyLight 350], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-88928UV
Product Code. 30307353
  • 737.00€ / 0.10mL
Description
myosin X Monoclonal antibody specifically detects myosin X in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
myosin X
Monoclonal
DyLight 350
FLJ10639, FLJ21066, FLJ22268, KIAA0799FLJ43256, MGC131988, myosin X, myosin-X, Unconventional myosin-10
This antibody was developed using a synthetic peptide derived from Q9HD67, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cell Biology, Cytoskeleton Markers, Signal Transduction
4651
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
CL9032
50mM Sodium Borate
Mouse
Protein A purified