missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYPT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22117-1-AP-20UL
This item is not returnable.
View return policy
Description
MYPT1, Myosin phosphatase target subunit 1, which is also called the myosin-binding subunit of myosin phosphatase, is one of the subunits of myosin phosphatase. Myosin phosphatase regulates the interaction of actin and myosin downstream of the guanosine triphosphatase Rho. The small guanosine triphosphatase Rho is implicated in myosin light chain (MLC) phosphorylation, which results in contraction of smooth muscle and interaction of actin and myosin in nonmuscle cells. The guanosine triphosphate (GTP)-bound, active form of RhoA (GTP.RhoA) specifically interacted with the myosin-binding subunit (MBS) of myosin phosphatase, which regulates the extent of phosphorylation of MLC. Rho-associated kinase (Rho-kinase), which is activated by GTP. RhoA, phosphorylated MBS and consequently inactivated myosin phosphatase. Overexpression of RhoA or activated RhoA in NIH 3T3 cells increased phosphorylation of MBS and MLC. Thus, Rho appears to inhibit myosin phosphatase through the action of Rho-kinase. Several transcript variants encoding different isoforms have been found for this gene.Specifications
| MYPT1 | |
| Polyclonal | |
| Unconjugated | |
| PPP1R12A | |
| MBS, MYPT1, PPP1R12A | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 116670, 17931, 4659 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Immunofluorescence, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| 0.64 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O14974, Q10728, Q9DBR7 | |
| Ppp1r12a | |
| MYPT1 Fusion Protein Ag17496 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction