missing translation for 'onlineSavingsMsg'
Learn More

N-(1-hydroxy-1-phenylpropan-2-yl)-N-methylformamide, TRC

Product Code. 30136015
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136015 5 mg 5mg
30008214 50 mg 50mg
30007765 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136015 Supplier Toronto Research Chemicals Supplier No. TRCC1031185MG
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91246-94-5
Molecular Formula C11H15NO2
Molecular Weight (g/mol) 193.24
Synonym N-(1-hydroxy-1-phenylpropan-2-yl)-N-methylformamide, rel-N-[(1R,2S)-2-Hydroxy-1-methyl-2-phenylethyl]-N-methylformamide
IUPAC Name N-(1-hydroxy-1-phenylpropan-2-yl)-N-methylformamide
SMILES CC(C(C1=CC=CC=C1)O)N(C)C=O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C11H15NO2/c1-9(12(2)8-13)11(14)10-6-4-3-5-7-10/h3-9,11,14H,1-2H3
Formula Weight 193.1103
Chemical Name or Material N-Formyl Ephedrine (Mixture of Diastereomers)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.