missing translation for 'onlineSavingsMsg'
Learn More

N-(1-Pyrenyl) Maleimide, TRC

Product Code. 30413816
Change view
Click to view available options
Quantity:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30413816 50 mg
30418138 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30413816 Supplier Toronto Research Chemicals Supplier No. TRCP99020050MG
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42189-56-0
Molecular Formula C20 H11 N O2
Molecular Weight (g/mol) 297.31
IUPAC Name 1-pyren-1-ylpyrrole-2,5-dione
SMILES O=C1C=CC(=O)N1c2ccc3ccc4cccc5ccc2c3c45

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C20H11NO2/c22-17-10-11-18(23)21(17)16-9-7-14-5-4-12-2-1-3-13-6-8-15(16)20(14)19(12)13/h1-11H
Formula Weight 297.079
Chemical Name or Material N-(1-Pyrenyl) Maleimide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.