Learn More
N-(10-Aminodecyl)-5-chloro-1-naphthalenesulfonamide Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | N-(10-Aminodecyl)-5-chloro-1-naphthalenesulfonamide hydrochloride |
| CAS | 79127-24-5 |
| Molecular Formula | C20 H29 Cl N2 O2 S . Cl H |
| Quantity | 100 mg |
| InChI Formula | InChI=1S/C20H29ClN2O2S.ClH/c21-19-13-9-12-18-17(19)11-10-14-20(18)26(24,25)23-16-8-6-4-2-1-3-5-7-15-22;/h9-14,23H,1-8,15-16,22H2;1H |
| SMILES | Cl.NCCCCCCCCCCNS(=O)(=O)c1cccc2c(Cl)cccc12 |
| IUPAC Name | N-(10-aminodecyl)-5-chloronaphthalene-1-sulfonamide;hydrochloride |
| Molecular Weight (g/mol) | 433.44 |
| Formula Weight | 432.1405 |