missing translation for 'onlineSavingsMsg'
Learn More

N-[(1S)-1-(Hydroxymethyl)-2-[(3S)-2-oxo-3-pyrrolidinyl]ethyl]-carbamic Acid tert-Butyl Ester, TRC

Product Code. 30399715
Change view
Click to view available options
Quantity:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30399715 250 mg
30407436 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30399715 Supplier Toronto Research Chemicals Supplier No. TRCH947785250MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 249736-45-6
Molecular Formula C12H22N2O4
Molecular Weight (g/mol) 258.31
Synonym N-[(1S)-1-(Hydroxymethyl)-2-[(3S)-2-oxo-3-pyrrolidinyl]ethyl]-carbamic Acid 1,1-Dimethylethyl Ester
IUPAC Name tert-butyl N-[(2S)-1-hydroxy-3-[(3S)-2-oxopyrrolidin-3-yl]propan-2-yl]carbamate
SMILES CC(C)(C)OC(=O)N[C@H](CO)C[C@@H]1CCNC1=O

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C12H22N2O4/c1-12(2,3)18-11(17)14-9(7-15)6-8-4-5-13-10(8)16/h8-9,15H,4-7H2,1-3H3,(H,13,16)(H,14,17)/t8-,9-/m0/s1
Formula Weight 258.158
Chemical Name or Material N-[(1S)-1-(Hydroxymethyl)-2-[(3S)-2-oxo-3-pyrrolidinyl]ethyl]-carbamic Acid tert-Butyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.