missing translation for 'onlineSavingsMsg'
Learn More

N-[2-(3,4-Dihydro-1-oxo-2(1H)-isoquinolinyl)-2-oxoethyl]-N-formyl-cyclohexanecarboxamide, TRC

Product Code. 30136668
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136668 50 mg 50mg
30009955 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30136668 Supplier Toronto Research Chemicals Supplier No. TRCD45008050MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 125273-88-3
Molecular Formula C19 H22 N2 O4
Molecular Weight (g/mol) 342.39
Synonym N-[2-(3,4-dihydro-1-oxo-2(1H)-isoquinolinyl)-2-oxoethyl]-N-formyl-cyclohexanecarboxamide, Praziquantel RC C (USP)
IUPAC Name N-formyl-N-[2-oxo-2-(1-oxo-3,4-dihydroisoquinolin-2-yl)ethyl]cyclohexanecarboxamide
SMILES O=CN(CC(=O)N1CCc2ccccc2C1=O)C(=O)C3CCCCC3

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C19H22N2O4/c22-13-20(18(24)15-7-2-1-3-8-15)12-17(23)21-11-10-14-6-4-5-9-16(14)19(21)25/h4-6,9,13,15H,1-3,7-8,10-12H2
Formula Weight 342.158
Chemical Name or Material 2-(N-formylhexahydrohippuroyl)-1,2,3,4-tetrahydroisoquinolin-1-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.