missing translation for 'onlineSavingsMsg'
Learn More

N-[[2-[[[4-[[[(Hexyloxy)carbonyl]amino]carbonyl]phenyl]amino]methyl]-1-methyl-1H-benzimidazol-5-yl]carbonyl]-N-2-pyridinyl-beta-alanine Ethyl Ester, TRC

Product Code. 30132695
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
Unit Size:
250mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30132695 500 mg 500mg
18080740 50 mg 50mg
18001240 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132695 Supplier Toronto Research Chemicals Supplier No. TRCH298150500MG
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1408238-40-3
Molecular Formula C34 H40 N6 O6
Molecular Weight (g/mol) 628.72
Synonym β-Alanine, N-[[2-[[[4-[[[(hexyloxy)carbonyl]amino]carbonyl]phenyl]amino]methyl]-1-methyl-1H-benzimidazol-5-yl]carbonyl]-N-2-pyridinyl-, ethyl ester, ethyl N-[(2-{[(4-{[(hexyloxy)carbonyl]carbamoyl}phenyl)amino]methyl}-1-methyl-1H-benzimidazol-5-yl) carbonyl]-N-pyridin-2-yl-β-alaninate, Dabigatran Etexilate Amid Analogon
IUPAC Name ethyl 3-[[2-[[4-(hexoxycarbonylcarbamoyl)anilino]methyl]-1-methylbenzimidazole-5-carbonyl]-pyridin-2-ylamino]propanoate
SMILES CCCCCCOC(=O)NC(=O)c1ccc(NCc2nc3cc(ccc3n2C)C(=O)N(CCC(=O)OCC)c4ccccn4)cc1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C34H40N6O6/c1-4-6-7-10-21-46-34(44)38-32(42)24-12-15-26(16-13-24)36-23-30-37-27-22-25(14-17-28(27)39(30)3)33(43)40(20-18-31(41)45-5-2)29-11-8-9-19-35-29/h8-9,11-17,19,22,36H,4-7,10,18,20-21,23H2,1-3H3,(H,38,42,44)
Formula Weight 628.3009
Percent Purity >95%
Chemical Name or Material Dabigatran Etexilate Amid Analogon
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.