missing translation for 'onlineSavingsMsg'
Learn More

N-(2,6-Dichlorophenyl)aniline, TRC

Product Code. 30138091
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
Unit Size:
25g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138091 25 g 25g
17901015 5 g 5g
17901825 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138091 Supplier Toronto Research Chemicals Supplier No. TRCD43406025G
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15307-93-4
Molecular Formula C12 H9 Cl2 N
Molecular Weight (g/mol) 238.11
Synonym 2,6-Dichlorodiphenylamine, Benzenamine, 2,6-dichloro-N-phenyl-, Diphenylamine, 2,6-dichloro- (8CI), N-(2,6-Dichlorophenyl)aniline, N-Phenyl-2,6-;dichloroaniline
IUPAC Name 2,6-dichloro-N-phenylaniline
SMILES Clc1cccc(Cl)c1Nc2ccccc2

Specifications

Quantity 25 g
InChI Formula InChI=1S/C12H9Cl2N/c13-10-7-4-8-11(14)12(10)15-9-5-2-1-3-6-9/h1-8,15H
Formula Weight 237.0112
Chemical Name or Material 2,6-Dichlorodiphenylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.