Learn More
N-(2-Bromo-5-nitrophenyl)benzenesulfonamide, 97%, Thermo Scientific™
Product Code. 15492546
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. 15492546
Brand: Thermo Scientific Alfa Aesar H56210.MD
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1421261-66-6
Molecular Formula C12H9BrN2O4S
Molecular Weight (g/mol) 357.18
MDL Number MFCD22682850
InChI Key KTKONBLIZGUGBA-UHFFFAOYSA-N
Synonym n-2-bromo-5-nitrophenyl benzenesulfonamide
PubChem CID 73996217
IUPAC Name N-(2-bromo-5-nitrophenyl)benzenesulfonamide
SMILES [O-][N+](=O)C1=CC(NS(=O)(=O)C2=CC=CC=C2)=C(Br)C=C1
Specifications
Quantity 250 mg
Formula Weight 357.18
Percent Purity 97%
Chemical Name or Material N-(2-Bromo-5-nitrophenyl)benzenesulfonamide