missing translation for 'onlineSavingsMsg'
Learn More

N-[(2-Chloro-5-thiazolyl)methyl]-N’-nitroguanidine, TRC

Product Code. 30138619
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
Unit Size:
250mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138619 500 mg 500mg
30008892 250 mg 250mg
30009293 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138619 Supplier Toronto Research Chemicals Supplier No. TRCC597070500MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 135018-15-4
Molecular Formula C5 H6 Cl N5 O2 S
Molecular Weight (g/mol) 235.65
Synonym N-[(2-Chloro-5-thiazolyl)methyl]-N'-nitroguanidine, CGA 265307, TZNG
IUPAC Name 1-[(2-chloro-1,3-thiazol-5-yl)methyl]-3-nitroguanidine
SMILES [O-][N+](=O)NC(=N)NCc1cnc(Cl)s1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C5H6ClN5O2S/c6-4-8-1-3(14-4)2-9-5(7)10-11(12)13/h1H,2H2,(H3,7,9,10)
Formula Weight 234.9931
Chemical Name or Material Clothianidin-desmethyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.