missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Chlorophenyl)maleimide, 97%, Thermo Scientific™

Product Code. 15529673 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15529673 1 g 1g
15575236 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15529673 Supplier Thermo Scientific Supplier No. 457330010
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 1203-24-3
Molecular Formula C10H6ClNO2
Molecular Weight (g/mol) 207.62
InChI Key KPQOXMCRYWDRSB-UHFFFAOYSA-N
Synonym 1-2-chlorophenyl-1h-pyrrole-2,5-dione, n-2-chlorophenyl maleimide, o-chphm, o-chlorophenylmaleimide, n-o-chlorophenylmaleimide, 1-2-chlorophenyl pyrrole-2,5-dione, 1h-pyrrole-2,5-dione, 1-2-chlorophenyl, n-2-chlorophenyl-maleimide, maleimide, n-o-chlorophenyl, n-o-chlorophenyl maleiimide
PubChem CID 14550
IUPAC Name 1-(2-chlorophenyl)pyrrole-2,5-dione
SMILES C1=CC=C(C(=C1)N2C(=O)C=CC2=O)Cl

Specifications

Melting Point 108.0°C to 109.5°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 207.62
Percent Purity 97%
Chemical Name or Material N-(2-Chlorophenyl)maleimide
compliance-icons
Product Identifier
  • N-(2-Chlorophenyl)maleimide, 97%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.