missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Hydroxyethyl)phthalimide, TRC

Product Code. 30404224
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30404224 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30404224 Supplier Toronto Research Chemicals Supplier No. TRCH949490250G
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-(2-Hydroxyethyl)phthalimide
CAS 3891-07-4
Molecular Formula C10H9NO3
Quantity 250 g
Synonym 2-(2-Hydroxyethyl)-1H-isoindole-1,3(2H)-dione, 2-(2-Hydroxyethyl)isoindole-1,3-dione, 2-(2-Hydroxyethyl)isoindoline-1,3-dione, 2-(N-Phthalimidyl)ethyl Alcohol, 2-Phthalimidoethanol, N-Hydroxyethylphthalimide, NSC 2773, NSC 50624
InChI Formula InChI=1S/C10H9NO3/c12-6-5-11-9(13)7-3-1-2-4-8(7)10(11)14/h1-4,12H,5-6H2
SMILES OCCN1C(=O)c2ccccc2C1=O
IUPAC Name 2-(2-hydroxyethyl)isoindole-1,3-dione
Molecular Weight (g/mol) 191.18
Formula Weight 191.0582
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.